Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172357.1 | NP_001165828.1 | 2 | 348 | ||
NM_001172410.1 | NP_001165881.1 | 2 | 348 | |||
NM_001317778.1 | NP_001304707.1 | 2 | 348 | |||
NM_001317780.1 | NP_001304709.1 | 2 | 348 | |||
NM_003018.3 | NP_003009.2 | 2 | 348 | |||
GenBank mRNA | AK058094.1 | - | 5 | 1040 | ||
AK298119.1 | - | 2 | 219 | |||
AK315742.1 | - | 2 | 222 | |||
BC005913.1 | - | 2 | 221 | 4043169 | ||
BG535994.1 | - | 2 | 225 | |||
BG569816.1 | - | 2 | 225 | |||
BI524154.1 | - | 2 | 205 | |||
DQ884411.1 | - | 2 | 190 | |||
J03517.1 | - | 2 | 217 | |||
J03553.1 | - | 2 | 364 | |||
KU178329.1 | - | 2 | 190 | |||
KU178330.1 | - | 2 | 190 |