Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277204.1 | NP_001264133.1 | 3 | 519 | ||
NM_001277205.1 | NP_001264134.1 | 4 | 466 | |||
NM_001277206.1 | NP_001264135.1 | 3 | 424 | |||
NM_001277207.1 | NP_001264136.1 | 3 | 366 | |||
NM_001277208.1 | NP_001264137.1 | 2 | 433 | |||
NM_176796.2 | NP_789766.1 | 4 | 499 | |||
NM_176797.2 | NP_789767.1 | 3 | 339 | |||
NM_176798.2 | NP_789768.1 | 4 | 616 | |||
XM_005274022.3 | XP_005274079.1 | 2 | 2607 | |||
XM_006718571.3 | XP_006718634.1 | 3 | 2692 | |||
XM_011545076.2 | XP_011543378.1 | 3 | 1822 | |||
XM_011545077.2 | XP_011543379.1 | 4 | 1941 | |||
XM_011545079.2 | XP_011543381.1 | 2 | 255 | |||
GenBank mRNA | AF007891.1 | - | 3 | 305 | ||
AF007892.1 | - | 4 | 616 | |||
AF007893.1 | - | 3 | 1139 | |||
AF218005.1 | - | 3 | 376 | |||
AF498920.1 | - | 1 | 74 | |||
AL520218.3 | - | 4 | 487 | |||
AL525099.3 | - | 4 | 461 | |||
AX775903.1 | - | 4 | 616 | |||
BC000571.2 | - | 2 | 314 | 3162734 | ||
BC009391.2 | - | 2 | 373 | 4128941 | ||
BQ718871.1 | - | 3 | 444 | |||
BT006771.1 | - | 1 | 74 | |||
BX337204.2 | - | 3 | 366 | |||
BX381745.2 | - | 4 | 487 | |||
BX397649.2 | - | 3 | 413 | |||
DR006998.1 | - | 4 | 588 | |||
U52464.1 | - | 3 | 277 | |||
X97058.1 | - | 3 | 350 |
Set Membership: |
G-protein coupled receptor |