Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198909.1 | NP_001185838.1 | 1 | 191 | ||
NM_004231.3 | NP_004222.2 | 1 | 191 | |||
GenBank mRNA | AK312214.1 | - | 1 | 136 | ||
BC104230.1 | - | 1 | 142 | 40032234 | ||
BC104231.1 | - | 1 | 142 | 40032235 | ||
BC107854.1 | - | 1 | 168 | 5171984 | ||
BG724326.1 | - | 1 | 176 | |||
BU956696.1 | - | 1 | 90 | |||
CR456896.1 | - | 1 | 89 | |||
D49400.1 | - | 1 | 139 | |||
DB499437.1 | - | 1 | 191 |