Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039574.2 | NP_001034663.1 | 1 | 902 | ||
NM_004978.4 | NP_004969.2 | 1 | 902 | |||
NR_036437.1 | 1 | 902 | ||||
XM_006710625.3 | XP_006710688.1 | 1 | 746 | |||
XM_006710627.3 | XP_006710690.1 | 1 | 746 | |||
XM_011541401.2 | XP_011539703.1 | 1 | 746 | |||
XM_011541402.1 | XP_011539704.1 | 1 | 746 | |||
XM_011541403.1 | XP_011539705.1 | 1 | 746 | |||
XM_011541404.1 | XP_011539706.1 | 1 | 746 | |||
GenBank mRNA | AK310193.1 | - | 1 | 327 | ||
BC101769.1 | - | 1 | 173 | 8069275 | ||
M64676.1 | - | 1 | 272 |