Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005185.3 | NP_005176.1 | 1 | 433 | ||
GenBank mRNA | AK313934.1 | - | 1 | 361 | ||
BC031889.1 | - | 1 | 341 | 4777524 | ||
HY060230.1 | - | 1 | 434 | |||
M36707.1 | - | 1 | 1301 | |||
M58026.1 | - | 1 | 361 |