Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005275.3 | NP_005266.2 | 5, 6 | 2000 | ||
XM_005249015.3 | XP_005249072.1 | 6, 7 | 1463 | |||
GenBank mRNA | AK026391.1 | - | 5, 6 | 662 | ||
AK298042.1 | - | 5, 6 | 839 | |||
AK301738.1 | - | 5, 6 | 590 | |||
AK302480.1 | - | 4, 5 | 728 | |||
AK316391.1 | - | 5, 6 | 601 | |||
BC013959.2 | - | 5, 6 | 649 | 2961417 | ||
BC018366.1 | - | 5, 6 | 1580 | 3924808 | ||
BC063657.1 | - | 2, 3 | 232 | |||
BT006648.1 | - | 5, 6 | 609 | |||
L25665.1 | - | 5, 6 | 559 |