Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAALADL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005468.2 | NP_005459.2 | 1, 2 | 201 | ||
XM_011544706.2 | XP_011543008.1 | 1, 2 | 521 | |||
XM_011544707.2 | XP_011543009.1 | 2, 3 | 169 | |||
XM_011544708.2 | XP_011543010.1 | 1, 2 | 521 | |||
XM_011544709.2 | XP_011543011.1 | 1, 2 | 541 | |||
XM_011544710.2 | XP_011543012.1 | 1, 2 | 538 | |||
XM_011544711.2 | XP_011543013.1 | 1, 2 | 543 | |||
XM_011544712.2 | XP_011543014.1 | 1, 2 | 544 | |||
GenBank mRNA | AJ012371.1 | - | 1, 2 | 201 |