Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006194.3 | NP_006185.1 | 3 | 1076 | ||
GenBank mRNA | AB248958.1 | - | 2 | 519 | ||
AK313627.1 | - | 2 | 531 | |||
BC001159.1 | - | 2 | 539 | 3534582 | ||
U59628.1 | - | 1 | 210 | |||
X92850.1 | - | 1 | 326 |