Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006402.2 | NP_006393.2 | 2 | 385 | ||
GenBank mRNA | AF029890.1 | - | 2 | 136 | ||
AK054769.1 | - | 1 | 1361 | |||
AY623819.1 | - | 2 | 81 | |||
BC062619.1 | - | 2 | 403 | 5190406 | ||
BX092018.1 | - | 2 | 401 | |||
CR456866.1 | - | 2 | 81 | |||
CR542130.1 | - | 2 | 81 |