Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006476.4 | NP_006467.4 | 2 | 357 | ||
NR_033759.1 | 3 | 410 | ||||
GenBank mRNA | AA258888.1 | - | 2 | 357 | ||
AF070655.1 | - | 2 | 141 | |||
AF087846.1 | - | 2 | 135 | |||
AF092124.1 | - | 2 | 153 | |||
AK289568.1 | - | 2 | 127 | |||
AL050277.1 | - | 2 | 139 | |||
BC015128.1 | - | 2 | 145 | 3930553 | ||
BC070165.1 | - | 2 | 114 | 6714804 | ||
BU953217.1 | - | 3 | 195 | |||
CR533494.1 | - | 2 | 80 | |||
CR542211.1 | - | 2 | 80 |