Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318217.1 | NP_001305146.1 | 4 | 431 | ||
NM_014187.3 | NP_054906.2 | 4 | 314 | |||
NR_134524.1 | 4 | 314 | ||||
GenBank mRNA | AF161519.1 | - | 4 | 316 | ||
BC003080.2 | - | 4 | 287 | 3050277 | ||
BC013412.2 | - | 4 | 266 | 3030307 | ||
BC021109.1 | - | 4 | 270 | 3660106 | ||
BF316569.1 | - | 4 | 240 | |||
BG825054.1 | - | 4 | 392 | |||
BI833541.1 | - | 4 | 273 |