Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318807.1 | NP_001305736.1 | 5, 6 | 1467 | ||
NM_014430.2 | NP_055245.2 | 4, 5 | 1318 | |||
XM_011536659.2 | XP_011534961.1 | 4, 5 | 974 | |||
XM_017021221.1 | XP_016876710.1 | 4, 5 | 3022 | |||
GenBank mRNA | AF190901.1 | - | 4, 5 | 1362 | ||
AF218586.1 | - | 2, 3 | 496 | |||
AF544398.1 | - | 2, 3 | 279 | |||
AK000387.1 | - | 2, 3 | 273 | |||
AK122828.1 | - | 4, 5 | 2910 | |||
BC035970.1 | - | 2, 3 | 306 | 4716269 | ||
BX418750.2 | - | 4, 5 | 873 | |||
CB162472.1 | - | 2, 3 | 254 |