Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303012.1 | NP_001289941.1 | 21 | 2755 | ||
NM_001303013.1 | NP_001289942.1 | 21 | 3941 | |||
NM_014638.3 | NP_055453.2 | 21 | 2949 | |||
XM_011542455.2 | XP_011540757.1 | 22 | 2991 | |||
XM_011542456.2 | XP_011540758.1 | 22 | 2948 | |||
XM_017002870.1 | XP_016858359.1 | 22 | 2980 | |||
XM_017002871.1 | XP_016858360.1 | 22 | 2951 | |||
XM_017002872.1 | XP_016858361.1 | 22 | 2980 | |||
XM_017002873.1 | XP_016858362.1 | 21 | 2872 | |||
XM_017002874.1 | XP_016858363.1 | 22 | 2980 | |||
GenBank mRNA | AB007919.2 | - | 21 | 2755 | ||
AK074149.1 | - | 10 | 1011 | |||
AK122591.2 | - | 8 | 894 | |||
BC019679.1 | - | 7 | 577 | |||
BC043358.1 | - | 18 | 2143 | |||
BC044630.1 | - | 4 | 303 | |||
BC128207.1 | - | 19 | 2331 | |||
BC144620.1 | - | 21 | 3933 | |||
DQ176850.1 | - | 21 | 2949 |