Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286102.1 | NP_001273031.1 | 2 | 260 | ||
NM_003269.4 | NP_003260.1 | 2 | 853 | |||
GenBank mRNA | AB307709.1 | - | 2 | 145 | ||
AF220532.1 | - | 2 | 852 | |||
AF411525.1 | - | 2 | 386 | |||
AK131541.1 | - | 2 | 260 | |||
BC028031.1 | - | 2 | 350 | 5242079 | ||
HQ692846.1 | - | 2 | 145 | |||
Y13276.1 | - | 2 | 313 |