Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102653.1 | NP_001096123.1 | 4 | 261 | ||
NM_017493.6 | NP_059963.1 | 3 | 159 | |||
XM_005263079.4 | XP_005263136.1 | 4 | 364 | |||
XM_011532040.2 | XP_011530342.2 | 4 | 365 | |||
XM_011532041.2 | XP_011530343.2 | 4 | 365 | |||
XM_011532042.2 | XP_011530344.1 | 4 | 286 | |||
GenBank mRNA | AK302581.1 | - | 4 | 223 | ||
BC118572.1 | - | 3 | 155 | 40036249 | ||
BC118653.1 | - | 3 | 155 | 40036248 | ||
BM471766.1 | - | 1 | 14 | |||
DB446926.2 | - | 4 | 263 | |||
X68242.1 | - | 4 | 250 |