Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104925.1 | NP_001098395.1 | 6 | 3262 | ||
NM_018257.2 | NP_060727.2 | 6 | 3343 | |||
GenBank mRNA | AK001745.1 | - | 6 | 3272 | ||
AK074436.1 | - | 1 | 2360 | |||
AK126700.1 | - | 2 | 3186 | |||
BC033665.2 | - | 5 | 3168 | 4999114 |