Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077238.1 | NP_001070706.1 | 2, 3 | 267 | ||
NM_152988.2 | NP_694533.1 | 2, 3 | 267 | |||
XM_011528138.2 | XP_011526440.1 | 2, 3 | 326 | |||
XM_011528139.1 | XP_011526441.1 | 1, 2 | 121 | |||
XM_017027016.1 | XP_016882505.1 | 2, 3 | 326 | |||
XM_017027018.1 | XP_016882507.1 | 1, 2 | 121 | |||
GenBank mRNA | AB040965.1 | - | 2, 3 | 216 | ||
AJ345027.1 | - | 2, 3 | 220 | |||
AJ420897.1 | - | 2, 3 | 216 | |||
AK308973.1 | - | 3, 4 | 336 | |||
AK309122.1 | - | 2, 3 | 341 | |||
AY169315.1 | - | 2, 3 | 215 | |||
BC001788.1 | - | 2, 3 | 201 | 3352341 | ||
BC028391.2 | - | 2, 3 | 204 | 4822524 | ||
BC093046.1 | - | 2, 3 | 267 | 30395602 |