Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024308.3 | NP_077284.2 | 2 | 557 | ||
XM_005257658.2 | XP_005257715.1 | 2 | 562 | |||
GenBank mRNA | AK026196.1 | - | 2 | 348 | ||
AK315735.1 | - | 2 | 478 | |||
AY358712.1 | - | 2 | 454 | |||
BC002731.2 | - | 2 | 443 | 3631177 | ||
CR457356.1 | - | 1 | 106 | |||
LC110386.1 | - | 2 | 343 |