Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46359
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
7
901
NM_001272105.1
NP_001259034.1
8
973
NM_001272107.1
NP_001259036.1
6
1036
NM_001272108.1
NP_001259037.1
5
526
NM_001272109.1
NP_001259038.1
4
791
NM_001272110.1
NP_001259039.1
6
774
NM_001272111.1
NP_001259040.1
6
732
NM_001272112.1
NP_001259041.1
4
559
NM_001272113.1
NP_001259042.1
4
429
NM_024897.3
NP_079173.2
6
1036
NM_198406.2
NP_940798.1
7
859
NR_073610.1
6
783
XM_005245494.3
XP_005245551.1
5
1027
XM_006711546.2
XP_006711609.1
7
799
XM_006711547.3
XP_006711610.1
7
1272
XM_006711548.3
XP_006711611.1
6
1154
XM_006711552.2
XP_006711615.1
5
897
XM_006711553.3
XP_006711616.1
5
939
XM_011510000.2
XP_011508302.1
6
1133
XM_017002385.1
XP_016857874.1
6
2430
GenBank mRNA
AF455045.1
-
6
955
AF455046.1
-
4
736
AF455047.1
-
6
697
AK026325.1
-
3
299
AK054682.1
-
6
713
AK094346.1
-
6
752
AK095143.1
-
7
760
AK095197.1
-
6
971
AK294143.1
-
6
711
AK295915.1
-
6
725
AK307441.1
-
5
563
AK316034.1
-
8
912
AY424284.1
-
7
780
BC058509.1
-
7
743
5770270
BI551776.1
-
6
675
BI819919.1
-
4
479
BP208594.1
-
5
464
BP362806.1
-
4
361
FJ200490.1
-
3
228
FJ200491.1
-
4
290
FJ200492.1
-
4
355
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline