Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160372.3 | NP_001153844.1 | 21 | 3135 | ||
NM_001321646.1 | NP_001308575.1 | 20 | 3108 | |||
NM_031466.7 | NP_113654.4 | 21 | 3310 | |||
XM_011517326.2 | XP_011515628.1 | 20 | 3283 | |||
XM_011517328.2 | XP_011515630.1 | 21 | 3310 | |||
XM_011517330.2 | XP_011515632.1 | 13 | 1675 | |||
XM_017013894.1 | XP_016869383.1 | 14 | 2011 | |||
GenBank mRNA | AB067469.1 | - | 21 | 3085 | ||
AK026394.1 | - | 21 | 3127 | |||
AK027689.1 | - | 10 | 1235 | |||
AK128755.1 | - | 21 | 3311 | |||
AK308815.1 | - | 10 | 1406 | |||
AL833973.1 | - | 19 | 2410 | |||
AY190606.1 | - | 19 | 2389 | |||
AY630619.1 | - | 2 | 190 | |||
BC003650.2 | - | 7 | 854 | |||
BC006206.2 | - | 19 | 2531 | 3538806 | ||
BC065288.1 | - | 4 | 395 |