Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032242.3 | NP_115618.3 | 31 | 8123 | ||
XM_011512908.2 | XP_011511210.1 | 32 | 8225 | |||
XM_011512909.1 | XP_011511211.1 | 32 | 8818 | |||
GenBank mRNA | AK054675.1 | - | 1 | 1251 | ||
AK127254.1 | - | 1 | 3295 | |||
AK128612.1 | - | 5 | 3260 | |||
AL136663.1 | - | 1 | 469 | |||
BC032432.1 | - | 1 | 309 |