Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006944.1 | NP_001006945.1 | 17 | 3077 | ||
NM_001300802.1 | NP_001287731.1 | 17 | 3074 | |||
NM_001318361.1 | NP_001305290.1 | 17 | 3049 | |||
NM_003942.2 | NP_003933.1 | 17 | 3095 | |||
XM_005274380.1 | XP_005274437.1 | 16 | 2910 | |||
XM_017018527.1 | XP_016874016.1 | 13 | 2495 | |||
GenBank mRNA | AJ010119.1 | - | 17 | 3077 | ||
AK223561.1 | - | 17 | 3079 | |||
BC047896.1 | - | 13 | 2330 | 5763859 |