Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252006.1 | NP_001238935.1 | 2 | 324 | ||
NM_001252007.1 | NP_001238936.1 | 2 | 324 | |||
NM_001293627.1 | NP_001280556.1 | 2 | 324 | |||
NM_001293628.1 | NP_001280557.1 | 2 | 189 | |||
NM_001293632.1 | NP_001280561.1 | 2 | 324 | |||
NM_032632.4 | NP_116021.2 | 2 | 324 | |||
XM_005267281.1 | XP_005267338.1 | 2 | 320 | |||
XM_005267282.2 | XP_005267339.1 | 2 | 320 | |||
XM_017020935.1 | XP_016876424.1 | 2 | 320 | |||
XM_017020936.1 | XP_016876425.1 | 2 | 320 | |||
GenBank mRNA | AK295020.1 | - | 2 | 155 | ||
AK302406.1 | - | 3 | 185 | |||
AK307659.1 | - | 2 | 283 | |||
BC000927.1 | - | 2 | 275 | |||
BC036014.1 | - | 2 | 319 | 5271668 | ||
BX161482.1 | - | 2 | 259 | |||
BX248301.1 | - | 2 | 189 | |||
BX248753.1 | - | 2 | 299 | |||
DB045100.1 | - | 2 | 325 | |||
DB483858.1 | - | 2 | 327 | |||
X76770.1 | - | 1 | 83 |