Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054032.3 | NP_473373.2 | 1 | 1322 | ||
GenBank mRNA | BC095509.1 | - | 1 | 1332 | 30706615 | |
BC107099.1 | - | 1 | 902 | 40015896 | ||
BC107100.1 | - | 1 | 902 | 40015904 |
Set Membership: |
G-protein coupled receptor |