Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017767.2 | NP_060237.2 | 1, 2 | 729 | ||
NM_130849.3 | NP_570901.2 | 2, 3 | 585 | |||
GenBank mRNA | AK000334.1 | - | 1, 2 | 709 | ||
AK000695.1 | - | 1, 2 | 646 | |||
AK025537.1 | - | 2, 3 | 579 | |||
AK290563.1 | - | 2, 3 | 533 | |||
BC062625.1 | - | 2, 3 | 565 | 5203858 | ||
KU178707.1 | - | 2, 3 | 479 |