Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170779.1 | NP_001164250.1 | 3 | 638 | ||
NM_138819.3 | NP_620174.1 | 3 | 638 | |||
XM_005262382.1 | XP_005262439.1 | 4 | 416 | |||
XM_005262383.2 | XP_005262440.1 | 4 | 416 | |||
XM_005262384.4 | XP_005262441.1 | 4 | 416 | |||
XM_005262386.3 | XP_005262443.1 | 4 | 416 | |||
XM_005262387.3 | XP_005262444.1 | 4 | 416 | |||
XM_006724734.3 | XP_006724797.1 | 4 | 504 | |||
XM_006724735.1 | XP_006724798.1 | 4 | 416 | |||
XM_006724736.2 | XP_006724799.1 | 4 | 416 | |||
XM_011531295.1 | XP_011529597.1 | 4 | 416 | |||
XM_011531296.2 | XP_011529598.1 | 4 | 416 | |||
XM_011531297.2 | XP_011529599.1 | 4 | 416 | |||
XM_011531298.2 | XP_011529600.1 | 4 | 416 | |||
XM_011531305.2 | XP_011529607.1 | 4 | 416 | |||
XM_011531306.2 | XP_011529608.1 | 4 | 416 | |||
XM_017029308.1 | XP_016884797.1 | 4 | 416 | |||
XM_017029311.1 | XP_016884800.1 | 4 | 494 | |||
GenBank mRNA | AK295485.1 | - | 4 | 396 | ||
BC017868.1 | - | 3 | 637 | 4699951 | ||
BC063491.1 | - | 3 | 578 | 5229041 |