Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165977.2 | NP_001159449.1 | 1 | 371 | ||
NM_001165978.2 | NP_001159450.1 | 1 | 371 | |||
NM_001321070.1 | NP_001307999.1 | 1 | 371 | |||
NM_144707.3 | NP_653308.2 | 1 | 371 | |||
XM_011510672.2 | XP_011508974.1 | 1 | 355 | |||
GenBank mRNA | AF245303.1 | - | 1 | 366 | ||
AF245304.1 | - | 1 | 366 | |||
AK091175.1 | - | 1 | 341 | |||
AK290366.1 | - | 1 | 326 | |||
AK313267.1 | - | 1 | 327 | |||
AY358377.1 | - | 1 | 322 | |||
BC113877.1 | - | 1 | 346 | |||
BC114525.1 | - | 1 | 346 | 40054105 | ||
DB003233.1 | - | 1 | 355 | |||
HY060111.1 | - | 1 | 372 |