Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145652.3 | NP_663627.1 | 2 | 251 | ||
XM_011528601.1 | XP_011526903.1 | 3 | 752 | |||
XM_011528602.1 | XP_011526904.1 | 4 | 560 | |||
GenBank mRNA | AY038181.1 | - | 2 | 211 | ||
AY358822.1 | - | 2 | 251 | |||
BC039173.1 | - | 2 | 217 | 4755260 |