Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF133 SIRNA Assays ›
Gene Symbol:
ZNF133
Gene Name:
zinc finger protein 133
Gene Aliases:
ZNF150, pHZ-13, pHZ-66
Chromosome Location:
Chr.20: 18288228 - 18316996 on Build GRCh38
UniGene ID:
Hs.472221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4652
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF133
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083330.2
NP_001076799.1
4
937
NM_001282995.1
NP_001269924.1
6
1173
NM_001282996.1
NP_001269925.1
6
1047
NM_001282997.1
NP_001269926.1
5
1098
NM_001282998.1
NP_001269927.1
4
927
NM_001282999.1
NP_001269928.1
4
940
NM_001283000.1
NP_001269929.1
5
1116
NM_001283001.1
NP_001269930.1
5
1103
NM_001283002.1
NP_001269931.1
5
1005
NM_001283003.1
NP_001269932.1
5
1078
NM_001283004.1
NP_001269933.1
5
1065
NM_001283005.1
NP_001269934.1
2
701
NM_001283006.1
NP_001269935.1
3
779
NM_001283007.1
NP_001269936.1
3
797
NM_001283008.1
NP_001269937.1
4
967
NM_003434.5
NP_003425.2
7
1349
XM_005260819.1
XP_005260876.1
4
749
XM_005260820.4
XP_005260877.1
6
1006
XM_011529336.1
XP_011527638.1
7
1085
XM_011529337.1
XP_011527639.1
6
957
XM_011529338.2
XP_011527640.1
6
951
XM_011529339.1
XP_011527641.1
7
1085
XM_011529340.1
XP_011527642.1
2
560
XM_017028041.1
XP_016883530.1
5
8850
XM_017028042.1
XP_016883531.1
5
2124
XM_017028043.1
XP_016883532.1
5
8847
XM_017028044.1
XP_016883533.1
7
9197
XM_017028045.1
XP_016883534.1
5
9265
XM_017028046.1
XP_016883535.1
5
8863
XM_017028047.1
XP_016883536.1
5
2217
XM_017028048.1
XP_016883537.1
4
2046
XM_017028049.1
XP_016883538.1
7
9207
XM_017028050.1
XP_016883539.1
7
1105
XM_017028051.1
XP_016883540.1
8
1281
XM_017028052.1
XP_016883541.1
6
945
XM_017028053.1
XP_016883542.1
4
979
XM_017028054.1
XP_016883543.1
5
859
XM_017028055.1
XP_016883544.1
8
1275
XM_017028056.1
XP_016883545.1
10
1494
XM_017028057.1
XP_016883546.1
7
1062
XM_017028058.1
XP_016883547.1
8
1279
GenBank mRNA
AK123005.1
-
2
2515
AK291389.1
-
5
855
AK294263.1
-
3
528
AK295273.1
-
5
777
AK295513.1
-
5
841
AK313525.1
-
4
676
AK316386.1
-
2
455
BC001887.1
-
7
1155
3535533
BP329060.1
-
3
555
BT007310.1
-
3
596
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline