Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206838.1 | NP_001193767.1 | 22 | 3692 | ||
NM_001206839.1 | NP_001193768.1 | 21 | 3671 | |||
NM_002851.2 | NP_002842.2 | 22 | 6272 | |||
XM_005250519.1 | XP_005250576.1 | 21 | 6251 | |||
XM_017012477.1 | XP_016867966.1 | 23 | 6319 | |||
GenBank mRNA | AK294062.1 | - | 11 | 1526 | ||
M93426.1 | - | 22 | 6021 | |||
X54135.1 | - | 11 | 1440 |