Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166347.1 | NP_001159819.1 | 3 | 455 | ||
NM_001166348.1 | NP_001159820.1 | 2 | 385 | |||
NM_001166349.1 | NP_001159821.1 | 2 | 255 | |||
NM_173626.3 | NP_775897.3 | 3 | 452 | |||
XM_006721833.1 | XP_006721896.1 | 3 | 455 | |||
XM_011524652.1 | XP_011522954.1 | 2 | 359 | |||
XM_011524653.1 | XP_011522955.1 | 2 | 359 | |||
XM_011524654.1 | XP_011522956.1 | 2 | 244 | |||
XM_017024505.1 | XP_016879994.1 | 2 | 369 | |||
XM_017024506.1 | XP_016879995.1 | 2 | 369 | |||
XM_017024507.1 | XP_016879996.1 | 3 | 429 | |||
GenBank mRNA | AF345195.1 | - | 2 | 187 | ||
AJ544073.1 | - | 3 | 426 | |||
AK027293.1 | - | 2 | 850 | |||
AK075248.1 | - | 3 | 415 | |||
AK315132.1 | - | 3 | 418 | |||
AL833468.1 | - | 2 | 349 | |||
BC035900.1 | - | 3 | 394 | 5241643 | ||
DA290933.1 | - | 3 | 457 |