Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012720.1 | NP_001012738.1 | 7 | 1147 | ||
NM_001012722.1 | NP_001012740.1 | 6 | 1033 | |||
NM_002921.3 | NP_002912.2 | 7 | 1159 | |||
GenBank mRNA | BC011349.1 | - | 7 | 1133 | 4157985 | |
BX641033.1 | - | 6 | 2321 | |||
U14910.1 | - | 7 | 1147 |
Set Membership: |
G-protein coupled receptor |