Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019376.2 | NP_062249.1 | 1, 2 | 237 | ||
XM_006249184.2 | XP_006249246.1 | 1, 2 | 365 | |||
GenBank mRNA | BC127496.1 | - | 1, 2 | 237 | 8372306 | |
D17447.1 | - | 1, 2 | 200 | |||
S55305.1 | - | 1, 2 | 200 |