Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022920.1 | NP_075209.1 | 1 | 337 | ||
XM_006246225.2 | XP_006246287.1 | 1 | 461 | |||
GenBank mRNA | D13963.1 | - | 1 | 337 |
Set Membership: |
![]() |