Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022945.1 | NP_075234.1 | 3 | 295 | ||
GenBank mRNA | X52016.1 | - | 3 | 295 |