Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sts | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012661.1 | NP_036793.1 | 5 | 1081 | ||
XM_006256960.2 | XP_006257022.1 | 5 | 1328 | |||
XM_006256961.2 | XP_006257023.1 | 5 | 1316 | |||
GenBank mRNA | BC085747.1 | - | 3 | 1419 | ||
U37138.1 | - | 5 | 1081 |