Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013154.2 | NP_037286.1 | 1 | 787 | ||
GenBank mRNA | M65149.1 | - | 1 | 756 |