Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022643.1 | NP_072169.1 | 1 | 234 | ||
GenBank mRNA | M18045.1 | - | 1 | 234 | ||
M18046.1 | - | 1 | 234 |