Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024377.1 | NP_077353.1 | 2 | 219 | ||
GenBank mRNA | BC058469.1 | - | 2 | 303 | 6918279 | |
DQ120493.1 | - | 2 | 164 | |||
DQ120494.1 | - | 2 | 164 | |||
M95780.1 | - | 2 | 219 |