Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012880.1 | NP_037012.1 | 2 | 1689 | ||
XM_006251030.2 | XP_006251092.1 | 2 | 1740 | |||
GenBank mRNA | BC061861.1 | - | 2 | 1645 | 5598594 | |
Z24721.1 | - | 2 | 1689 |