Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003105.5 | NP_003096.1 | 42 | 5840 | ||
XM_011542963.2 | XP_011541265.1 | 41 | 5601 | |||
XM_011542965.2 | XP_011541267.1 | 36 | 4749 | |||
XM_011542967.2 | XP_011541269.1 | 22 | 2781 | |||
XM_017018169.1 | XP_016873658.1 | 42 | 5547 | |||
XM_017018170.1 | XP_016873659.1 | 40 | 5223 | |||
XM_017018172.1 | XP_016873661.1 | 28 | 3712 | |||
GenBank mRNA | AK294404.1 | - | 13 | 1978 | ||
AK295090.1 | - | 19 | 2630 | |||
AK295878.1 | - | 22 | 2781 | |||
AK302589.1 | - | 19 | 2452 | |||
AK316340.1 | - | 13 | 1560 | |||
BC137171.1 | - | 42 | 5763 | 9021168 | ||
Y08110.1 | - | 42 | 5791 |