Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019150.1 | NP_062023.1 | 2 | 401 | ||
GenBank mRNA | U33935.1 | - | 2 | 401 |