Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024141.1 | NP_077055.1 | 28 | 3794 | ||
XM_006234862.2 | XP_006234924.1 | 30 | 4758 | |||
GenBank mRNA | AF237962.1 | - | 28 | 3794 | ||
AF547268.1 | - | 6 | 803 |