Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031833.2 | NP_114021.2 | 2, 3 | 296 | ||
GenBank mRNA | BC086599.1 | - | 2, 3 | 296 | 7312852 | |
FQ217120.1 | - | 1, 2 | 197 | |||
FQ217563.1 | - | 2, 3 | 183 | |||
FQ218179.1 | - | 1, 2 | 213 | |||
FQ222529.1 | - | 2, 3 | 181 | |||
M55331.1 | - | 1, 2 | 153 | |||
M91597.1 | - | 2, 3 | 187 |