Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270564.1 | NP_001257493.1 | 1 | 549 | ||
NM_001270565.1 | NP_001257494.1 | 1 | 549 | |||
NM_001270566.1 | NP_001257495.1 | 1 | 549 | |||
NM_001270567.1 | NP_001257496.1 | 1 | 549 | |||
NM_001270568.1 | NP_001257497.1 | 1 | 549 | |||
NM_001270569.1 | NP_001257498.1 | 1 | 549 | |||
NM_001270570.1 | NP_001257499.1 | 1 | 549 | |||
NM_053506.2 | NP_445958.1 | 1 | 549 | |||
GenBank mRNA | AB015646.1 | - | 1 | 549 | ||
AF237919.1 | - | 1 | 212 | |||
AY009370.1 | - | 1 | 212 | |||
AY009371.1 | - | 1 | 212 | |||
BC087707.1 | - | 1 | 547 | 7315203 | ||
DQ112342.1 | - | 1 | 212 | |||
DQ112343.1 | - | 1 | 212 | |||
FQ212445.1 | - | 1 | 532 |
Set Membership: |
G-protein coupled receptor |