Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053724.3 | NP_446176.3 | 111 | |||
GenBank mRNA | AJ310838.1 | - | 111 | |||
M55250.1 | - | 462 |
Gene Symbol: LOC100909864 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771239.1 | XP_008769461.1 | 1, 2 | 556 |