Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003168.2 | NP_003159.1 | 5 | 1313 | ||
NR_073470.1 | 5 | 1315 | ||||
GenBank mRNA | AK129758.1 | - | 5 | 942 | ||
BG026473.1 | - | 1 | 478 | |||
U38818.1 | - | 5 | 1271 | |||
U43923.1 | - | 5 | 1277 |