Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138826.4 | NP_620181.1 | 3 | 265 | ||
GenBank mRNA | AF411318.1 | - | 1 | 99 | ||
BC058442.1 | - | 3 | 270 | 6891030 | ||
DQ255899.1 | - | 1 | 67 | |||
FQ209933.1 | - | 3 | 266 | |||
FQ210006.1 | - | 3 | 266 | |||
FQ210086.1 | - | 3 | 266 | |||
FQ210225.1 | - | 3 | 266 | |||
FQ210276.1 | - | 3 | 266 | |||
FQ210404.1 | - | 3 | 266 | |||
FQ210574.1 | - | 3 | 266 | |||
FQ210578.1 | - | 3 | 266 | |||
FQ210592.1 | - | 3 | 266 | |||
FQ210741.1 | - | 3 | 266 | |||
FQ210753.1 | - | 3 | 266 | |||
FQ210805.1 | - | 3 | 266 | |||
FQ210924.1 | - | 3 | 266 | |||
FQ210951.1 | - | 3 | 266 | |||
FQ211146.1 | - | 3 | 266 | |||
FQ211312.1 | - | 3 | 266 | |||
FQ211350.1 | - | 3 | 266 | |||
FQ217792.1 | - | 3 | 266 | |||
FQ218077.1 | - | 3 | 266 | |||
FQ218157.1 | - | 3 | 266 | |||
FQ218315.1 | - | 3 | 266 | |||
FQ220411.1 | - | 3 | 266 | |||
FQ221163.1 | - | 3 | 266 | |||
FQ221173.1 | - | 3 | 266 | |||
FQ221343.1 | - | 3 | 266 | |||
FQ221498.1 | - | 3 | 266 | |||
FQ221507.1 | - | 3 | 266 | |||
FQ221511.1 | - | 3 | 266 | |||
FQ221516.1 | - | 3 | 266 | |||
FQ221571.1 | - | 3 | 266 | |||
FQ221639.1 | - | 3 | 266 | |||
FQ221768.1 | - | 3 | 266 | |||
FQ221874.1 | - | 3 | 266 | |||
FQ222065.1 | - | 3 | 266 | |||
FQ222070.1 | - | 3 | 266 | |||
FQ222149.1 | - | 3 | 266 | |||
FQ222220.1 | - | 3 | 266 | |||
FQ222329.1 | - | 3 | 266 | |||
FQ222341.1 | - | 3 | 266 | |||
FQ222400.1 | - | 3 | 266 | |||
FQ222420.1 | - | 3 | 266 | |||
FQ222436.1 | - | 3 | 266 | |||
FQ222448.1 | - | 3 | 266 | |||
FQ222605.1 | - | 3 | 266 | |||
FQ222614.1 | - | 3 | 265 | |||
FQ222637.1 | - | 3 | 266 | |||
FQ222674.1 | - | 3 | 266 | |||
FQ222691.1 | - | 3 | 266 | |||
FQ222786.1 | - | 3 | 266 | |||
FQ222845.1 | - | 3 | 266 | |||
FQ222891.1 | - | 3 | 266 | |||
FQ222907.1 | - | 3 | 266 | |||
FQ222910.1 | - | 3 | 266 | |||
FQ222929.1 | - | 3 | 266 | |||
FQ222950.1 | - | 3 | 266 | |||
FQ222957.1 | - | 3 | 265 | |||
FQ223018.1 | - | 3 | 266 | |||
FQ223045.1 | - | 3 | 266 | |||
FQ223097.1 | - | 3 | 266 | |||
FQ223170.1 | - | 3 | 266 | |||
FQ223179.1 | - | 3 | 266 | |||
FQ223313.1 | - | 3 | 266 | |||
FQ223425.1 | - | 3 | 266 | |||
FQ223452.1 | - | 3 | 266 | |||
FQ228316.1 | - | 3 | 266 | |||
FQ228334.1 | - | 3 | 266 | |||
FQ228356.1 | - | 3 | 266 | |||
FQ228476.1 | - | 3 | 266 | |||
FQ228561.1 | - | 3 | 266 | |||
FQ228573.1 | - | 3 | 266 | |||
FQ228587.1 | - | 3 | 266 | |||
FQ228764.1 | - | 3 | 266 | |||
FQ228920.1 | - | 3 | 266 | |||
FQ228980.1 | - | 3 | 266 | |||
FQ229826.1 | - | 3 | 266 | |||
FQ229982.1 | - | 3 | 266 | |||
J00750.1 | - | 3 | 265 | |||
M24327.1 | - | 1 | 69 |
Gene Symbol: Ttr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ210386.1 | - | 266 |