Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138892.1 | NP_620247.1 | 2, 3 | 259 | ||
GenBank mRNA | AF331839.1 | - | 2, 3 | 233 | ||
AY029335.1 | - | 2, 3 | 259 |