Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdipt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138899.2 | NP_620254.1 | 2 | 562 | ||
XM_006230208.1 | XP_006230270.1 | 2 | 736 | |||
GenBank mRNA | BC070876.1 | - | 2 | 562 | 7096385 | |
D82928.1 | - | 2 | 310 |